Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017031.2 | NP_058727.2 | 5, 6 | 1100 | ||
GenBank mRNA | AF202732.1 | - | 958 | |||
AF202733.1 | - | 5, 6 | 664 | |||
J04563.1 | - | 2, 3 | 112 | |||
U95748.1 | - | 5, 6 | 1100 |