Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113328.1 | NP_001106799.1 | 5 | 770 | ||
NM_001113329.1 | NP_001106800.1 | 6 | 685 | |||
NM_001113332.1 | NP_001106803.1 | 5 | 622 | |||
NM_001113334.1 | NP_001106805.1 | 5 | 459 | |||
NM_017032.1 | NP_058728.1 | 5 | 377 | |||
XM_008760721.1 | XP_008758943.1 | 5 | 1778 | |||
GenBank mRNA | AF031373.1 | - | 5 | 770 | ||
AF536979.1 | - | 6 | 685 | |||
AY388961.1 | - | 5 | 395 | |||
EF102484.1 | - | 5 | 622 | |||
EF121818.1 | - | 5 | 459 | |||
L27059.1 | - | 2 | 96 | |||
U09457.1 | - | 5 | 377 |