Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017033.2 | NP_058729.2 | 3 | 520 | ||
XM_006238442.2 | XP_006238504.1 | 4 | 711 | |||
XM_008763853.1 | XP_008762075.1 | 3 | 1063 | |||
GenBank mRNA | BC099807.1 | - | 3 | 520 | 7937943 | |
BC128703.1 | - | 3 | 525 | |||
L11694.1 | - | 3 | 503 | |||
U20195.1 | - | 2 | 103 |