Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022519.2 | NP_071964.2 | 5 | 1241 | ||
XM_006240456.1 | XP_006240518.1 | 4 | 1098 | |||
GenBank mRNA | BC078824.1 | - | 5 | 1241 | 7131535 | |
D00675.1 | - | 4 | 1044 | |||
FQ209479.1 | - | 3 | 555 | |||
FQ209494.1 | - | 3 | 558 | |||
FQ209517.1 | - | 3 | 555 | |||
FQ209677.1 | - | 3 | 555 | |||
FQ209686.1 | - | 3 | 555 | |||
FQ209716.1 | - | 3 | 556 | |||
FQ209779.1 | - | 3 | 556 | |||
FQ209814.1 | - | 3 | 556 | |||
FQ209897.1 | - | 3 | 555 | |||
FQ209914.1 | - | 3 | 555 | |||
FQ210068.1 | - | 3 | 555 | |||
FQ210169.1 | - | 3 | 555 | |||
FQ210668.1 | - | 2 | 177 | |||
FQ210685.1 | - | 3 | 555 | |||
FQ210775.1 | - | 3 | 555 | |||
FQ210832.1 | - | 3 | 475 | |||
FQ210989.1 | - | 3 | 556 | |||
FQ210996.1 | - | 3 | 556 | |||
FQ211012.1 | - | 3 | 555 | |||
FQ211042.1 | - | 3 | 555 | |||
FQ211203.1 | - | 3 | 555 | |||
FQ211353.1 | - | 3 | 555 | |||
FQ211455.1 | - | 3 | 421 | |||
FQ218226.1 | - | 3 | 555 | |||
FQ218241.1 | - | 3 | 555 | |||
FQ218252.1 | - | 3 | 556 | |||
FQ218261.1 | - | 3 | 555 | |||
FQ218411.1 | - | 4 | 1174 | |||
FQ218460.1 | - | 3 | 555 | |||
FQ218555.1 | - | 3 | 555 | |||
FQ218567.1 | - | 3 | 555 | |||
FQ218586.1 | - | 3 | 555 | |||
FQ218654.1 | - | 3 | 555 | |||
FQ218694.1 | - | 3 | 556 | |||
FQ218710.1 | - | 3 | 554 | |||
FQ218855.1 | - | 3 | 555 | |||
FQ218857.1 | - | 3 | 556 | |||
FQ218947.1 | - | 3 | 556 | |||
FQ218955.1 | - | 3 | 556 | |||
FQ218993.1 | - | 3 | 555 | |||
FQ219224.1 | - | 3 | 555 | |||
FQ219268.1 | - | 3 | 555 | |||
FQ219276.1 | - | 3 | 555 | |||
FQ219303.1 | - | 3 | 555 | |||
FQ219383.1 | - | 3 | 555 | |||
FQ219441.1 | - | 3 | 555 | |||
FQ219458.1 | - | 3 | 555 | |||
FQ219621.1 | - | 3 | 555 | |||
FQ219652.1 | - | 3 | 555 | |||
FQ219732.1 | - | 3 | 555 | |||
FQ219741.1 | - | 4 | 1035 | |||
FQ219765.1 | - | 3 | 555 | |||
M32247.1 | - | 3 | 981 | |||
X16273.1 | - | 3 | 427 |