Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166356.1 | NP_001159828.1 | 7 | 1011 | ||
NM_001166357.1 | NP_001159829.1 | 7 | 1103 | |||
NM_001166358.1 | NP_001159830.1 | 7 | 1002 | |||
NM_001166359.1 | NP_001159831.1 | 7 | 895 | |||
NM_005412.5 | NP_005403.2 | 7 | 1041 | |||
NR_029415.1 | 6 | 843 | ||||
NR_029416.1 | 6 | 922 | ||||
NR_029417.1 | 6 | 961 | ||||
NR_048562.1 | 6 | 1051 | ||||
XM_011538675.2 | XP_011536977.1 | 7 | 981 | |||
XM_011538676.2 | XP_011536978.1 | 7 | 972 | |||
XM_011538677.2 | XP_011536979.1 | 6 | 882 | |||
XM_011538678.2 | XP_011536980.1 | 6 | 864 | |||
GenBank mRNA | AK055053.1 | - | 4 | 1691 | ||
AK223555.1 | - | 7 | 903 | |||
AK295941.1 | - | 6 | 792 | |||
AK296183.1 | - | 7 | 855 | |||
AK297173.1 | - | 6 | 803 | |||
AK298231.1 | - | 6 | 696 | |||
AK301339.1 | - | 7 | 768 | |||
AK301443.1 | - | 6 | 865 | |||
AK303827.1 | - | 6 | 717 | |||
AK315916.1 | - | 7 | 1095 | |||
BC011911.2 | - | 7 | 845 | 4139001 | ||
BC013677.2 | - | 7 | 905 | 3863368 | ||
BC025355.1 | - | 4 | 428 | |||
BC032584.1 | - | 7 | 884 | 5499481 | ||
BC044211.1 | - | 7 | 881 | 5521082 | ||
BC091501.1 | - | 6 | 765 | |||
BT006866.1 | - | 7 | 835 | |||
BX647711.1 | - | 7 | 962 | |||
BX648847.1 | - | 5 | 443 | |||
EU668344.1 | - | 7 | 845 | |||
KU178333.1 | - | 7 | 835 | |||
KU178334.1 | - | 6 | 471 | |||
L11932.1 | - | 6 | 746 |