Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017042.2 | NP_058738.1 | 13 | 1499 | ||
GenBank mRNA | D90036.1 | - | 13 | 1499 | ||
FQ214219.1 | - | 545 | ||||
M31809.1 | - | 13 | 1499 |