Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012634.1 | NP_036766.1 | 4 | 783 | ||
XM_006256829.2 | XP_006256891.1 | 4 | 555 | |||
GenBank mRNA | M17259.1 | - | 4 | 524 | ||
M29393.1 | - | 4 | 524 | |||
X16555.1 | - | 4 | 783 |
Gene Symbol: LOC100910245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256848.2 | XP_006256910.2 | 4 | 633 | ||
XM_008773204.1 | XP_008771426.1 | 559 |