Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109887.1 | NP_001103357.1 | 7 | 508 | ||
NM_001109888.1 | NP_001103358.1 | 7 | 487 | |||
NM_001109889.1 | NP_001103359.1 | 7 | 484 | |||
NM_001109890.1 | NP_001103360.1 | 6 | 361 | |||
NM_138507.2 | NP_612516.2 | 9 | 757 | |||
XM_006249910.2 | XP_006249972.1 | 9 | 851 | |||
XM_006249912.2 | XP_006249974.1 | 8 | 685 | |||
XM_008769534.1 | XP_008767756.1 | 8 | 688 | |||
GenBank mRNA | AF251010.1 | - | 5 | 235 | ||
M10072.1 | - | 1 | 100 | |||
M25820.1 | - | 5 | 316 | |||
M25821.1 | - | 5 | 313 | |||
M25822.1 | - | 5 | 337 | |||
M25823.1 | - | 7 | 586 | |||
Y00065.1 | - | 7 | 586 |