Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001041.3 | NP_001032.2 | 27 | 3198 | ||
XM_011513078.2 | XP_011511380.1 | 27 | 3202 | |||
GenBank mRNA | BC115034.1 | - | 27 | 3211 | 7962025 | |
BC116452.1 | - | 27 | 3211 | 7962021 | ||
BC116453.1 | - | 27 | 3211 | |||
BC132834.1 | - | 27 | 3191 | 40146856 | ||
BC132860.1 | - | 27 | 3191 | 40146882 | ||
X63597.1 | - | 27 | 3197 |