Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ret | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110099.1 | NP_001103569.1 | 14, 15 | 2631 | ||
NM_012643.2 | NP_036775.2 | 14, 15 | 2631 | |||
GenBank mRNA | AJ299016.1 | - | 14, 15 | 2634 | ||
AJ299017.1 | - | 14, 15 | 2634 |