Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031530.1 | NP_113718.1 | 3 | 363 | ||
GenBank mRNA | AF058786.1 | - | 3 | 363 | ||
M57441.1 | - | 3 | 327 |