Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003032.2 | NP_003023.1 | 5 | 1069 | ||
NM_173216.2 | NP_775323.1 | 6 | 1392 | |||
NM_173217.2 | NP_775324.1 | 5 | 735 | |||
XM_005247719.1 | XP_005247776.1 | 5 | 980 | |||
XM_005247720.1 | XP_005247777.1 | 5 | 1038 | |||
XM_006713734.1 | XP_006713797.1 | 7 | 1647 | |||
XM_011513085.1 | XP_011511387.1 | 7 | 1655 | |||
XM_011513086.1 | XP_011511388.1 | 5 | 1038 | |||
XM_017007065.1 | XP_016862554.1 | 8 | 1739 | |||
XM_017007066.1 | XP_016862555.1 | 5 | 1067 | |||
XM_017007067.1 | XP_016862556.1 | 5 | 1187 | |||
XM_017007068.1 | XP_016862557.1 | 5 | 1072 | |||
GenBank mRNA | A17362.1 | - | 6 | 1196 | ||
A23699.1 | - | 3 | 773 | |||
AK128726.1 | - | 1 | 929 | |||
AK292879.1 | - | 5 | 1090 | |||
AK312023.1 | - | 2 | 98 | |||
BC031476.1 | - | 5 | 1063 | 4339124 | ||
BC040009.1 | - | 5 | 1069 | 5557868 | ||
DB235666.1 | - | 5 | 524 | |||
X17247.1 | - | 6 | 1190 | |||
X54363.1 | - | 5 | 1028 | |||
X62822.1 | - | 5 | 1069 |