Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012652.1 | NP_036784.1 | 1, 2 | 1119 | ||
GenBank mRNA | M85299.1 | - | 1, 2 | 1119 |