Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPARC SIRNA Assays ›
Gene Symbol:
Sparc
Gene Name:
secreted protein, acidic, cysteine-rich (osteonectin)
Gene Aliases:
-
Chromosome Location:
Chr.10: 40742390 - 40764222 on Build Rnor_6.0
UniGene ID:
Rn.234316
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s128432
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sparc
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012656.1
NP_036788.1
10
868
XM_008767663.1
XP_008765885.1
9
946
GenBank mRNA
BC061777.1
-
9
883
5621666
D28875.1
-
9
885
FQ212364.1
-
9
937
FQ213815.1
-
9
902
FQ214222.1
-
9
900
FQ214492.1
-
9
902
FQ214649.1
-
9
903
FQ214673.1
-
9
902
FQ215195.1
-
9
901
FQ215581.1
-
9
903
FQ215622.1
-
9
904
FQ215818.1
-
9
913
FQ215935.1
-
9
902
FQ216823.1
-
9
904
FQ217546.1
-
9
903
FQ219866.1
-
9
884
FQ219932.1
-
10
884
FQ219983.1
-
9
902
FQ220003.1
-
9
902
FQ220043.1
-
9
900
FQ220046.1
-
9
902
FQ220186.1
-
9
900
FQ220279.1
-
7
726
FQ220287.1
-
9
904
FQ220443.1
-
9
884
FQ220507.1
-
9
910
FQ220547.1
-
9
902
FQ220572.1
-
9
902
FQ220644.1
-
9
902
FQ220880.1
-
9
902
FQ220901.1
-
9
902
FQ222224.1
-
9
902
FQ223025.1
-
9
902
FQ224996.1
-
8
773
FQ225033.1
-
9
904
FQ225092.1
-
9
898
FQ225200.1
-
9
902
FQ225220.1
-
9
881
FQ225486.1
-
9
884
FQ225609.1
-
9
902
FQ225618.1
-
9
900
FQ225644.1
-
5
569
FQ225678.1
-
9
902
FQ225688.1
-
9
902
FQ225694.1
-
9
904
FQ225722.1
-
9
901
FQ225884.1
-
9
900
FQ225913.1
-
9
902
FQ225914.1
-
9
904
FQ225965.1
-
9
902
FQ226299.1
-
9
884
FQ226354.1
-
9
902
FQ226360.1
-
9
902
FQ226584.1
-
9
902
FQ226599.1
-
9
903
FQ226651.1
-
9
925
FQ226673.1
-
9
884
FQ226731.1
-
9
902
FQ226746.1
-
9
900
FQ227279.1
-
9
903
FQ227628.1
-
9
911
FQ227644.1
-
9
902
FQ227708.1
-
9
903
FQ227725.1
-
9
903
FQ227727.1
-
9
882
FQ227749.1
-
9
902
FQ228032.1
-
9
882
FQ229172.1
-
9
902
FQ229199.1
-
9
902
FQ229230.1
-
9
902
FQ229283.1
-
9
884
FQ229299.1
-
9
885
FQ229315.1
-
9
900
FQ229316.1
-
9
902
FQ230029.1
-
9
902
FQ230086.1
-
9
900
FQ230210.1
-
9
903
FQ230381.1
-
9
902
FQ230595.1
-
9
884
FQ230667.1
-
9
903
FQ230705.1
-
9
900
FQ230778.1
-
9
902
FQ230864.1
-
9
902
FQ230993.1
-
9
902
FQ231067.1
-
9
902
FQ231166.1
-
9
901
FQ231444.1
-
9
901
FQ231523.1
-
9
902
FQ231543.1
-
9
902
FQ231632.1
-
9
902
FQ231753.1
-
9
899
FQ231815.1
-
9
884
FQ231918.1
-
9
884
FQ231975.1
-
9
884
FQ232042.1
-
9
901
FQ232191.1
-
9
902
FQ232223.1
-
9
902
FQ232271.1
-
9
899
FQ232364.1
-
9
904
FQ232445.1
-
9
900
FQ232464.1
-
9
902
FQ232710.1
-
9
900
FQ232838.1
-
9
902
FQ232847.1
-
9
902
FQ232862.1
-
9
900
FQ232869.1
-
9
902
FQ232965.1
-
9
902
FQ232977.1
-
9
902
FQ232986.1
-
9
902
FQ233178.1
-
9
902
FQ233283.1
-
9
881
FQ233669.1
-
9
902
FQ233698.1
-
9
884
FQ233777.1
-
9
900
FQ233810.1
-
9
904
FQ233845.1
-
9
902
FQ233850.1
-
9
903
FQ233926.1
-
9
902
FQ234069.1
-
9
902
FQ234108.1
-
9
902
FQ234199.1
-
9
902
FQ234410.1
-
9
884
FQ234418.1
-
9
902
FQ234523.1
-
9
889
FQ234589.1
-
9
900
FQ234657.1
-
9
884
FQ234685.1
-
9
902
FQ234766.1
-
9
900
FQ235008.1
-
9
904
FQ235038.1
-
9
903
FQ235049.1
-
9
884
Y13714.1
-
10
868
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix glycoprotein
Gene Ontology Categories:
Function(s)
Process(es)
ossification
negative regulation of endothelial cell proliferation
signal transduction
heart development
response to gravity
response to lead ion
positive regulation of endothelial cell migration
negative regulation of angiogenesis
regulation of cell morphogenesis
lung development
response to lipopolysaccharide
response to L-ascorbic acid
response to cytokine
wound healing
regulation of cell proliferation
response to peptide hormone
response to ethanol
response to cadmium ion
inner ear development
response to glucocorticoid
response to cAMP
response to calcium ion
bone development
cellular response to growth factor stimulus
calcium ion binding
collagen binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-zrdb5:80/100.66.79.80:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline