Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032056.3 | NP_114445.2 | 2 | 148 | ||
GenBank mRNA | BC091140.1 | - | 1 | 81 | 7381526 | |
KC222910.1 | - | 1 | 59 | |||
KC222911.1 | - | 1 | 59 | |||
X63854.1 | - | 2 | 148 | |||
X75305.1 | - | 2 | 95 | |||
X75306.1 | - | 2 | 95 | |||
X75307.1 | - | 2 | 95 |
Gene Symbol: LOC103689996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772777.1 | XP_008770999.1 | 2 | 200 |