Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012670.1 | NP_036802.1 | 2 | 213 | ||
GenBank mRNA | D90345.1 | - | 2 | 213 |