Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098845.2 | NP_001092315.2 | 3 | 390 | ||
NM_001278923.1 | NP_001265852.1 | 3 | 390 | |||
NM_001278924.1 | NP_001265853.1 | 3 | 504 | |||
GenBank mRNA | AK223149.1 | - | 3 | 204 | ||
AK300191.1 | - | 3 | 224 | |||
AK307182.1 | - | 3 | 204 | |||
AK313626.1 | - | 3 | 253 | |||
BC008813.2 | - | 3 | 418 | 3958020 | ||
DC381265.1 | - | 3 | 391 | |||
X16662.1 | - | 3 | 234 |
Gene Symbol: ANXA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040084.2 | NP_001035173.1 | 3 | 353 | ||
NM_001271702.1 | NP_001258631.1 | 3 | 467 | |||
XM_006717951.3 | XP_006718014.1 | 3 | 307 | |||
XM_011540096.1 | XP_011538398.1 | 4 | 506 | |||
XM_011540098.1 | XP_011538400.1 | 4 | 506 | |||
XM_011540099.1 | XP_011538401.1 | 4 | 506 | |||
XM_011540101.2 | XP_011538403.1 | 4 | 506 | |||
GenBank mRNA | AK296976.1 | - | 3 | 257 | ||
AK298761.1 | - | 3 | 333 | |||
AK300139.1 | - | 3 | 318 | |||
AK300158.1 | - | 3 | 211 | |||
BC004376.1 | - | 3 | 250 | 3639064 | ||
BC073755.1 | - | 3 | 284 | 6538697 | ||
CB270161.1 | - | 3 | 339 | |||
DC339657.1 | - | 3 | 353 | |||
M81844.1 | - | 3 | 155 | |||
Z36830.1 | - | 3 | 264 |