Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270854.1 | NP_001257783.1 | 7 | 748 | ||
NM_012672.3 | NP_036804.2 | 8 | 856 | |||
XM_006251709.2 | XP_006251771.1 | 3 | 769 | |||
XM_008770500.1 | XP_008768722.1 | 5 | 660 | |||
GenBank mRNA | DQ191165.1 | - | 5 | 603 | ||
HM043807.1 | - | 4 | 645 | |||
J03819.1 | - | 7 | 744 | |||
J03933.1 | - | 7 | 745 |