Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052800.1 | NP_434687.1 | 6 | 545 | ||
GenBank mRNA | CV072953.1 | - | 6 | 544 | ||
FQ229684.1 | - | 6 | 585 | |||
FQ234584.1 | - | 555 | ||||
FQ234769.1 | - | 555 |