Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022922.2 | NP_075211.2 | 1 | 107 | ||
GenBank mRNA | BC061781.1 | - | 1 | 82 | ||
BC126087.1 | - | 1 | 107 | 8368624 | ||
FQ214607.1 | - | 1 | 192 | |||
FQ214670.1 | - | 1 | 115 | |||
FQ214880.1 | - | 1 | 120 | |||
FQ215529.1 | - | 1 | 119 | |||
FQ215810.1 | - | 1 | 120 | |||
FQ216584.1 | - | 1 | 119 | |||
FQ223237.1 | - | 1 | 121 | |||
FQ224880.1 | - | 1 | 118 | |||
FQ224920.1 | - | 1 | 122 | |||
FQ224970.1 | - | 1 | 115 | |||
L36250.1 | - | 1 | 107 |
Gene Symbol: LOC100911515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750654.3 | XP_003750702.2 | 1 | 263 |