Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022922.2 | NP_075211.2 | 7 | 1218 | ||
GenBank mRNA | BC061781.1 | - | 7 | 1193 | ||
BC126087.1 | - | 7 | 1218 | 8368624 | ||
FQ215213.1 | - | 6 | 1133 | |||
FQ216226.1 | - | 6 | 1156 | |||
FQ216307.1 | - | 7 | 1117 | |||
FQ223237.1 | - | 7 | 1232 | |||
FQ224920.1 | - | 7 | 1233 | |||
L36250.1 | - | 7 | 1208 |
Gene Symbol: LOC100911515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750654.3 | XP_003750702.2 | 7 | 1374 |