Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022922.2 | NP_075211.2 | 3 | 314 | ||
GenBank mRNA | BC061781.1 | - | 3 | 289 | ||
BC126087.1 | - | 3 | 314 | 8368624 | ||
FQ214607.1 | - | 3 | 399 | |||
FQ214670.1 | - | 3 | 322 | |||
FQ214880.1 | - | 3 | 327 | |||
FQ215213.1 | - | 2 | 229 | |||
FQ215529.1 | - | 3 | 326 | |||
FQ215606.1 | - | 2 | 202 | |||
FQ215810.1 | - | 3 | 327 | |||
FQ216226.1 | - | 2 | 252 | |||
FQ216307.1 | - | 3 | 213 | |||
FQ216584.1 | - | 3 | 326 | |||
FQ223237.1 | - | 3 | 328 | |||
FQ224880.1 | - | 3 | 325 | |||
FQ224920.1 | - | 3 | 329 | |||
FQ224970.1 | - | 3 | 322 | |||
L36250.1 | - | 3 | 314 |
Gene Symbol: LOC100911515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750654.3 | XP_003750702.2 | 3 | 470 |