Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769997.1 | XP_008768219.1 | 5 | 1613 | ||
XM_008769998.1 | XP_008768220.1 | 5 | 2370 | |||
XM_008769999.1 | XP_008768221.1 | 5 | 1267 |
Gene Symbol: LOC100364155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770097.1 | XP_008768319.1 | 5 | 1326 |
Gene Symbol: Ugt2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031533.5 | NP_113721.4 | 5 | 1273 | ||
GenBank mRNA | BC089792.1 | - | 5 | 1256 | 7370057 | |
DQ351215.1 | - | 5 | 1614 | |||
X03478.1 | - | 5 | 1162 |