Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024823.4 | NP_001019994.4 | 3179 | |||
NM_001159653.1 | NP_001153125.1 | 1848 | ||||
NM_019132.1 | NP_062005.1 | 1108 | ||||
NM_021845.5 | NP_068617.6 | 3179 | ||||
GenBank mRNA | AF105254.1 | - | 1706 | |||
AF107844.1 | - | 1666 | ||||
BC061967.1 | - | 1070 | ||||
BC158589.1 | - | 963 | 9099119 | |||
BC168156.1 | - | 972 | ||||
DQ120475.1 | - | 957 | ||||
DQ120476.1 | - | 957 | ||||
FQ212511.1 | - | 935 | ||||
FQ214546.1 | - | 846 | ||||
FQ219804.1 | - | 538 | ||||
FQ226208.1 | - | 636 | ||||
M12673.1 | - | 1172 | ||||
M17525.1 | - | 1108 | ||||
X84047.1 | - | 2327 |