Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012697.1 | NP_036829.1 | 2 | 481 | ||
GenBank mRNA | X78855.1 | - | 2 | 481 |