Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031539.1 | NP_113727.1 | 3 | 481 | ||
GenBank mRNA | BC059127.1 | - | 3 | 503 | 6888323 | |
X04310.1 | - | 3 | 481 |