Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172091.2 | NP_742088.1 | 12 | 1197 | ||
NM_172092.2 | NP_742089.1 | 12 | 1205 | |||
XM_006247876.2 | XP_006247938.1 | 12 | 1311 | |||
XM_006247877.1 | XP_006247939.1 | 10 | 1036 | |||
XM_008768468.1 | XP_008766690.1 | 11 | 1254 | |||
XM_008768469.1 | XP_008766691.1 | 11 | 1257 | |||
GenBank mRNA | BC081700.1 | - | 11 | 1062 | 7192550 | |
CV117246.1 | - | 8 | 721 | |||
L04796.1 | - | 12 | 1167 | |||
M96674.1 | - | 12 | 1207 | |||
X68692.1 | - | 12 | 1167 |
Set Membership: |
G-protein coupled receptor |