Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008829.1 | NP_001008829.1 | 2 | 182 | ||
GenBank mRNA | BC088465.1 | - | 2 | 198 | 7127909 | |
FQ233418.1 | - | 2 | 208 | |||
X90376.1 | - | 1 | 116 |
Gene Symbol: RT1-EC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF400159.1 | - | 2 | 197 | ||
U38972.1 | - | 3 | 179 | |||
X82106.1 | - | 1 | 116 | |||
Y08532.1 | - | 2 | 116 |