Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-Cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206848.1 | NP_996730.1 | 4 | 487 | ||
GenBank mRNA | EU040040.1 | - | 2 | 424 |