Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080769.2 | NP_542947.1 | 4 | 456 |
Gene Symbol: LOC103690381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772785.1 | XP_008771007.1 | 1 | 89 |