Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1563136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_215875.5 | XP_215875.4 | 3 | 394 |
Gene Symbol: Andpro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012718.2 | NP_036850.1 | 3 | 446 | ||
XM_006235184.2 | XP_006235246.1 | 3 | 685 | |||
GenBank mRNA | BC065311.1 | - | 1 | 3866 | ||
M27901.1 | - | 3 | 430 | |||
M58167.1 | - | 3 | 447 |
Gene Symbol: P22k15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199266.2 | NP_954887.1 | 414 | |||
GenBank mRNA | BC100632.1 | - | 414 | 5621817 | ||
M58169.1 | - | 445 |