Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012721.2 | NP_036853.2 | 3 | 458 | ||
XM_008768851.1 | XP_008767073.1 | 3 | 648 | |||
XM_008768852.1 | XP_008767074.1 | 3 | 333 | |||
GenBank mRNA | X92070.1 | - | 3 | 339 | ||
X97376.1 | - | 3 | 469 |