Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012728.1 | NP_036860.1 | 7, 8 | 850 | ||
GenBank mRNA | M97797.1 | - | 7, 8 | 850 | ||
S75952.1 | - | 7, 8 | 844 |
Set Membership: |
G-protein coupled receptor |