Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001140.3 | NP_001131.3 | 7 | 834 | ||
GenBank mRNA | AK290309.1 | - | 7 | 847 | ||
AK296821.1 | - | 8 | 919 | |||
AK296904.1 | - | 7 | 730 | |||
AK312557.1 | - | 7 | 847 | |||
AK316126.1 | - | 7 | 990 | |||
BC029032.1 | - | 7 | 834 | 5179700 | ||
M23892.1 | - | 7 | 823 |