Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006930.3 | NP_008861.2 | 2 | 252 | ||
NM_170679.2 | NP_733779.1 | 2 | 252 | |||
GenBank mRNA | BC009839.2 | - | 2 | 136 | 4053593 | |
BC020798.1 | - | 2 | 187 | 4243711 | ||
BC025673.1 | - | 2 | 149 | 5203008 | ||
BC065730.1 | - | 2 | 173 | 6672613 | ||
BI552966.1 | - | 2 | 219 | |||
BI670085.1 | - | 2 | 257 | |||
U37558.1 | - | 1 | 34 | |||
Z47087.1 | - | 2 | 182 |