Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243120.1 | NP_001230049.1 | 5 | 845 | ||
NM_005983.3 | NP_005974.2 | 7 | 1092 | |||
NM_032637.3 | NP_116026.1 | 7 | 1092 | |||
XM_011514081.2 | XP_011512383.1 | 7 | 1146 | |||
XM_011514082.2 | XP_011512384.1 | 7 | 1146 | |||
XM_011514083.2 | XP_011512385.1 | 6 | 895 | |||
XM_017009753.1 | XP_016865242.1 | 6 | 895 | |||
GenBank mRNA | AB050979.1 | - | 6 | 872 | ||
AB050980.1 | - | 6 | 872 | |||
AB050981.1 | - | 6 | 872 | |||
AK291255.1 | - | 7 | 1024 | |||
AK295152.1 | - | 7 | 966 | |||
AK296223.1 | - | 5 | 760 | |||
AY029177.1 | - | 7 | 924 | |||
BC001441.2 | - | 7 | 1011 | 3138709 | ||
BC007441.2 | - | 7 | 1015 | 2962938 |