Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031543.1 | NP_113731.1 | 5 | 686 | ||
XM_006230520.1 | XP_006230582.1 | 6 | 869 | |||
GenBank mRNA | AF061442.1 | - | 5 | 674 | ||
BC081774.1 | - | 5 | 688 | 7110096 | ||
FQ209397.1 | - | 5 | 704 | |||
FQ210615.1 | - | 5 | 615 | |||
FQ210815.1 | - | 5 | 707 | |||
FQ211086.1 | - | 5 | 707 | |||
FQ212595.1 | - | 2 | 52 | |||
FQ218528.1 | - | 3 | 325 | |||
FQ218532.1 | - | 5 | 702 | |||
FQ219179.1 | - | 2 | 52 | |||
FQ219736.1 | - | 5 | 704 | |||
J02627.1 | - | 5 | 686 |