Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012742.1 | NP_036874.1 | 1 | 100 | ||
GenBank mRNA | X55955.1 | - | 1 | 100 |