Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012744.2 | NP_036876.2 | 14 | 2098 | ||
XM_006230689.2 | XP_006230751.1 | 15 | 2329 | |||
XM_006230690.2 | XP_006230752.1 | 14 | 2444 | |||
XM_006230691.2 | XP_006230753.1 | 15 | 2151 | |||
XM_006230692.2 | XP_006230754.1 | 14 | 2121 | |||
XM_006230693.2 | XP_006230755.1 | 15 | 2253 | |||
XM_006230694.2 | XP_006230756.1 | 14 | 2120 | |||
XM_008760087.1 | XP_008758309.1 | 14 | 2226 | |||
GenBank mRNA | BC085680.1 | - | 14 | 2075 | 7109369 | |
U32314.1 | - | 14 | 2066 | |||
U36585.1 | - | 14 | 2098 |