Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aanat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012818.2 | NP_036950.1 | 3, 4 | 419 | ||
XM_006247792.1 | XP_006247854.1 | 3, 4 | 1415 | |||
XM_006247793.1 | XP_006247855.1 | 3, 4 | 1410 | |||
GenBank mRNA | DQ075321.1 | - | 3, 4 | 607 | ||
U29664.1 | - | 2, 3 | 208 | |||
U38306.1 | - | 3, 4 | 419 | |||
U40803.1 | - | 3, 4 | 506 |