Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022380.1 | NP_071775.1 | 1371 | |||
GenBank mRNA | X91988.1 | - | 9, 10 | 1405 | ||
X97541.1 | - | 1371 |