Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017084.1 | NP_058780.1 | 4, 5 | 585 | ||
GenBank mRNA | X06150.1 | - | 4, 5 | 585 |
Gene Symbol: LOC100911564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766920.1 | XP_008765142.1 | 4, 5 | 617 |