Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022847.1 | NP_074038.1 | 8 | 2584 | ||
GenBank mRNA | HM037361.1 | - | 3136 | |||
HM037362.1 | - | 3136 | ||||
L16922.1 | - | 8 | 2584 | |||
S68283.1 | - | 2 | 191 | |||
S68285.1 | - | 2 | 191 |