Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031551.1 | NP_113739.1 | 5, 6 | 414 | ||
XM_006229551.1 | XP_006229613.1 | 3, 4 | 836 | |||
XM_006229552.2 | XP_006229614.1 | 4, 5 | 419 | |||
XM_006229553.2 | XP_006229615.1 | 5, 6 | 439 | |||
XM_006229554.1 | XP_006229616.1 | 4, 5 | 366 | |||
GenBank mRNA | BC083688.1 | - | 5, 6 | 414 | 7189278 | |
FQ214482.1 | - | 5, 6 | 380 | |||
FQ214526.1 | - | 5, 6 | 380 | |||
FQ214664.1 | - | 5, 6 | 355 | |||
FQ214718.1 | - | 5, 6 | 430 | |||
FQ214896.1 | - | 5, 6 | 365 | |||
FQ216691.1 | - | 5, 6 | 370 | |||
X74125.1 | - | 5, 6 | 340 |