Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012766.1 | NP_036898.1 | 2 | 332 | ||
XM_008766836.1 | XP_008765058.1 | 2 | 109 | |||
GenBank mRNA | BC085763.1 | - | 2 | 365 | 7189593 | |
BC089819.1 | - | 522 | 7384331 | |||
D16309.1 | - | 2 | 332 |