Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270711.1 | NP_001257640.1 | 10 | 1246 | ||
NM_012770.2 | NP_036902.2 | 8 | 1129 | |||
XM_008770737.1 | XP_008768959.1 | 10 | 1377 | |||
XM_008770738.1 | XP_008768960.1 | 10 | 1377 | |||
XM_008770739.1 | XP_008768961.1 | 9 | 1729 | |||
XM_008770740.1 | XP_008768962.1 | 9 | 1506 | |||
XM_008770741.1 | XP_008768963.1 | 10 | 1377 | |||
GenBank mRNA | AB058888.1 | - | 7 | 1004 | ||
AB109448.1 | - | 7 | 983 | |||
AB109449.1 | - | 7 | 983 | |||
AB109450.1 | - | 7 | 983 | |||
AB109451.1 | - | 7 | 983 | |||
AY004153.2 | - | 10 | 1246 | |||
M57507.1 | - | 8 | 1129 |