Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272087.1 | NP_001259016.1 | 8 | 2163 | ||
NM_001272088.1 | NP_001259017.1 | 6 | 991 | |||
NM_005071.2 | NP_005062.1 | 5 | 769 | |||
NR_073589.1 | 5 | 722 | ||||
XM_006722842.1 | XP_006722905.1 | 6 | 952 | |||
XM_006722844.1 | XP_006722907.1 | 6 | 845 | |||
XM_017027152.1 | XP_016882641.1 | 6 | 1040 | |||
GenBank mRNA | AK302380.1 | - | 5 | 699 | ||
AK312996.1 | - | 6 | 1035 | |||
BC028721.2 | - | 6 | 913 | 5269300 | ||
BC040604.1 | - | 8 | 2118 | 5270965 | ||
U18244.1 | - | 5 | 769 |