Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000340.1 | NP_000331.1 | 10, 11 | 1691 | ||
NM_001278658.1 | NP_001265587.1 | 9, 10 | 1428 | |||
NM_001278659.1 | NP_001265588.1 | 9, 10 | 1566 | |||
XM_011513087.2 | XP_011511389.1 | 11, 12 | 1438 | |||
XM_011513089.2 | XP_011511391.1 | 7, 8 | 979 | |||
GenBank mRNA | AK290846.1 | - | 10, 11 | 1434 | ||
AK292741.1 | - | 10, 11 | 1434 | |||
AK298418.1 | - | 9, 10 | 1192 | |||
AK313622.1 | - | 10, 11 | 1432 | |||
BC060041.1 | - | 9, 10 | 1311 | 4716156 | ||
J03810.1 | - | 10, 11 | 1420 |