Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001631.4 | NP_001622.2 | 3, 4 | 396 | ||
GenBank mRNA | AK313163.1 | - | 3, 4 | 385 | ||
BC132678.1 | - | 3, 4 | 338 | 40146700 | ||
DA376676.1 | - | 3, 4 | 396 | |||
DA925041.1 | - | 3, 4 | 385 | |||
M15694.1 | - | 3, 4 | 308 | |||
M31008.1 | - | 2, 3 | 398 |