Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000340.1 | NP_000331.1 | 7 | 1254 | ||
NM_001278658.1 | NP_001265587.1 | 6 | 991 | |||
NM_001278659.1 | NP_001265588.1 | 6 | 1129 | |||
XM_011513087.2 | XP_011511389.1 | 8 | 1001 | |||
XM_011513089.2 | XP_011511391.1 | 4 | 542 | |||
GenBank mRNA | AK290846.1 | - | 7 | 997 | ||
AK292741.1 | - | 7 | 997 | |||
AK298418.1 | - | 6 | 755 | |||
AK313622.1 | - | 7 | 995 | |||
BC060041.1 | - | 6 | 874 | 4716156 | ||
J03810.1 | - | 7 | 983 |